Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decyl methyl ether | 7289-52-3 | MFCD00216758 | >98.0% | 172.31 g/mol | C11H24O | 50 MG
SDP

Catalog No. 5000284611
Encompass_Preferred

Decyl methyl ether (1-methoxydecane) is a fatty ether supplied for research use. It is reported as non-genotoxic and can be phototoxic/photosensitizing. Supplied in small laboratory pack sizes; consult the certificate of analysis for lot-specific purity and storage instructions.

  • Also known as 1-methoxydecane.
  • CAS number 7289-52-3.
  • Molecular formula C11H24O.
  • Molecular weight 172.31 g/mol.
  • Purity ≥98.0% (lot-specific certificate of analysis).
  • Reported non-genotoxic and potentially phototoxic/photosensitizing.
  • Available in small research pack sizes (e.g., 25 mg, 50 mg, 100 mg); larger quantities available by quote.

Catalog No. 50-002-84611 Supplier Medchemexpress LLC Supplier No. HYW58789250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decyl methyl ether (1-methoxydecane) is a fatty ether supplied for research use. It is reported as non-genotoxic and can be phototoxic/photosensitizing. Supplied in small laboratory pack sizes; consult the certificate of analysis for lot-specific purity and storage instructions.

  • Also known as 1-methoxydecane.
  • CAS number 7289-52-3.
  • Molecular formula C11H24O.
  • Molecular weight 172.31 g/mol.
  • Purity ≥98.0% (lot-specific certificate of analysis).
  • Reported non-genotoxic and potentially phototoxic/photosensitizing.
  • Available in small research pack sizes (e.g., 25 mg, 50 mg, 100 mg); larger quantities available by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.