Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decylsuccinic Anhydride 90.0+%, TCI America™
SDP

Catalog No. D003625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D003625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D003625G Supplier TCI America Supplier No. D003625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18470-76-3
Molecular Formula C14H24O3
Molecular Weight (g/mol) 240.34
MDL Number MFCD00014548
InChI Key YOWKKGPNCDIFFB-UHFFFAOYNA-N
PubChem CID 86743
IUPAC Name 3-decyloxolane-2,5-dione
SMILES CCCCCCCCCCC1CC(=O)OC1=O

Specifications

Melting Point 69°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 240.34
Percent Purity ≥90.0% (T)
Chemical Name or Material Decylsuccinic Anhydride
RTECSNumber WN1080000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.