Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyltriethoxysilane 98.0+%, TCI America™
SDP

Catalog No. D519725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D519725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D519725ML Supplier TCI America Supplier No. D519725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2943-73-9
Molecular Formula C16H36O3Si
Molecular Weight (g/mol) 304.546
MDL Number MFCD00069132
InChI Key BAAAEEDPKUHLID-UHFFFAOYSA-N
Synonym n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy
PubChem CID 76261
IUPAC Name decyl(triethoxy)silane
SMILES CCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 150°C
Quantity 25 mL
Formula Weight 304.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyltriethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.