Learn More
Medchemexpress LLC Decyltrimethylammonium chloride | 10108-87-9 | ≥98.0% | 235.84 | 25 G
SDP
Catalog No. 5000451266
Encompass_Preferred
N,N,N-Trimethyldecan-1-aminium chloride is a quaternary ammonium compound used in various industrial and biomedical applications. It functions as a surfactant, emulsifier, and disinfectant. It is also used as a phase transfer catalyst in organic synthesis reactions to facilitate reactant movement between immiscible phases.
  • Salt with cationic properties
  • Used in personal care products, detergents, textiles, and pharmaceutical formulations
  • Facilitates reactant movement between immiscible phases
  • Biochemical reagent for life science-related research
Catalog No. 50-004-51266 Supplier Medchemexpress LLC Supplier No. 25GHYW012167
N,N,N-Trimethyldecan-1-aminium chloride is a quaternary ammonium compound used in various industrial and biomedical applications. It functions as a surfactant, emulsifier, and disinfectant. It is also used as a phase transfer catalyst in organic synthesis reactions to facilitate reactant movement between immiscible phases.
  • Salt with cationic properties
  • Used in personal care products, detergents, textiles, and pharmaceutical formulations
  • Facilitates reactant movement between immiscible phases
  • Biochemical reagent for life science-related research