Learn More
Enzo Life Sciences Decylubiquinone (10mg). CAS: 55486-00-5
SDP
Catalog No. 502010418
Encompass_Preferred
Inhibits the Ca2+ dependent mitochondrial permeability transition pore (MPTP) without inhibiting respiration. Maximum effect on Ca2+ retention capacity of rat liver mitochondria at 100μM. Inhibits etoposide-induced apoptosis in in L929 cells at 5μM. Alternative name: 2,3-Dimethoxy-5-methyl-6-decyl-1,4-benzoquinone. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0418 Supplier Enzo Life Sciences Supplier No. BMLCM1150010
Inhibits the Ca2+ dependent mitochondrial permeability transition pore (MPTP) without inhibiting respiration. Maximum effect on Ca2+ retention capacity of rat liver mitochondria at 100μM. Inhibits etoposide-induced apoptosis in in L929 cells at 5μM. Alternative name: 2,3-Dimethoxy-5-methyl-6-decyl-1,4-benzoquinone. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.