Learn More
Cayman Chemical Decylubiquinone, 55486-00-5, 50 mg
SDP
Supplier:  Cayman Chemical 2102750
Encompass_Preferred
An analog of ubiquinone that blocks opening of the mitochondrial permeability transition pore
Catalog No. NC1362941