Learn More
Enzo Life Sciences 7,7-Dimethyleicosadienoic acid (50mg). CAS: 89560-01-0
SDP
Catalog No. 502010556
Encompass_Preferred
Non-metabolizable analog of arachidonic or eicosadienoic acid. Cell permeable. DEDA inhibits P388D1 cell phospholipase A2 (PLA2) (IC50=16 µM) and snake venom PLA2 (IC50=14 µM). It inhibits human recombinant sPLA2 (IC50=5.8 µM) selectively over cPLA2. It is also a weak inhibitor of 5-lipoxygenase (IC50=56 µM, RBL-l cells). Inhibits induction of the anti-viral state in human fibroblasts induced by interferon. Alternative name: DEDA. Purity: ≥99%. Solubility: Soluble in 100% ethanol or methanol. Long Term Storage: +4°C.
Catalog No. 50-201-0556 Supplier Enzo Life Sciences Supplier No. BMLEI1120050
Non-metabolizable analog of arachidonic or eicosadienoic acid. Cell permeable. DEDA inhibits P388D1 cell phospholipase A2 (PLA2) (IC50=16 µM) and snake venom PLA2 (IC50=14 µM). It inhibits human recombinant sPLA2 (IC50=5.8 µM) selectively over cPLA2. It is also a weak inhibitor of 5-lipoxygenase (IC50=56 µM, RBL-l cells). Inhibits induction of the anti-viral state in human fibroblasts induced by interferon. Alternative name: DEDA. Purity: ≥99%. Solubility: Soluble in 100% ethanol or methanol. Long Term Storage: +4°C.