Learn More
IONBENCH CORPORATION Dedicated Mobile Lab Bench, 190 cm W x 88 cm D x 56 cm H, Integrated Pump Noise-Reduction Enclosure, Patented Vibration Dampening, Casters, Overheat Alarm, Tubing & Solvent Storage
Catalog No. NC3739848
Encompass_Preferred
Dedicated laboratory bench engineered for mass spectrometry and other sensitive analytical instruments, providing noise reduction, vibration isolation, mobility and integrated utility management to protect instrument performance and simplify routine maintenance.
  • Dimensions: 190 cm width x 88 cm depth x 56 cm height.
  • Integrated noise reduction enclosure for vacuum pumps to reduce ambient laboratory noise.
  • Patented vibration dampening system to prevent vibrations from reaching sensitive instruments.
  • Robust casters for full mobility and easy repositioning.
  • Audible and visual overheating alarm for the vacuum pump enclosure.
  • Integrated tubing and cabling management and dedicated solvent storage.
  • Quick and easy oil-change access for vacuum pump maintenance.
  • Catalog No. NC3739848 Supplier IONBENCH CORPORATION Supplier No. BCH190NECLH56
    Add to Cart
    Add to Cart
    Request a Quote
    Dedicated laboratory bench engineered for mass spectrometry and other sensitive analytical instruments, providing noise reduction, vibration isolation, mobility and integrated utility management to protect instrument performance and simplify routine maintenance.
  • Dimensions: 190 cm width x 88 cm depth x 56 cm height.
  • Integrated noise reduction enclosure for vacuum pumps to reduce ambient laboratory noise.
  • Patented vibration dampening system to prevent vibrations from reaching sensitive instruments.
  • Robust casters for full mobility and easy repositioning.
  • Audible and visual overheating alarm for the vacuum pump enclosure.
  • Integrated tubing and cabling management and dedicated solvent storage.
  • Quick and easy oil-change access for vacuum pump maintenance.
  • Product Title