Learn More
USA Scientific Inc 0.5 mL deep 96-well PP plate, sterile
Catalog No. 507541653
Encompass
0.5 mL deep 96-well sterile PlateOne polypropylene plate, round wells, natural, individually wrapped, 32/pack
Catalog No. 50-754-1653 Supplier USA Scientific Inc Supplier No. 18965110
0.5 mL deep 96-well sterile PlateOne polypropylene plate, round wells, natural, individually wrapped, 32/pack