Learn More
USA Scientific Inc 1 mL deep 96-well PP plate, bulk
Catalog No. NC1600748
Encompass
1 mL deep 96-well PlateOne polypropylene plate, round wells, natural, bulk 20/pack
Catalog No. NC1600748 Supplier USA Scientific Inc Supplier No. 18961000
1 mL deep 96-well PlateOne polypropylene plate, round wells, natural, bulk 20/pack