Learn More
GA International Deep-Freeze Cover-Up Labels for Thermal-Transfer Barcode Printers 3" x 1.25" / 76.2mm x 31.8mm Rectangle White From -80°C to +120°C (-112°F to +248°F)
Catalog No. 5000571845
Encompass_Preferred
Deep-freeze permanent blackout labels are designed to conceal any already existing information or labels. Ideal for re-labeling cryo vials, microtubes, and freezer boxes, to be stored in ultra-low temperature freezers (-80°C) or to be transported in dry ice.These cover-up labels will completely obscure the existing label and block-out the information underneath. Printable with all major brands of thermal-transfer printers, these versatile labels can withstand a wide temperature range, also suitable for high-temperature applications, as high as +120°C.
Catalog No. 50-005-71845 Supplier GA International Supplier No. BOC86C11WH
Deep-freeze permanent blackout labels are designed to conceal any already existing information or labels. Ideal for re-labeling cryo vials, microtubes, and freezer boxes, to be stored in ultra-low temperature freezers (-80°C) or to be transported in dry ice.These cover-up labels will completely obscure the existing label and block-out the information underneath. Printable with all major brands of thermal-transfer printers, these versatile labels can withstand a wide temperature range, also suitable for high-temperature applications, as high as +120°C.