Learn More
GA International Deep-Freeze Labels for High-Profile PCR Tube Strips 3" x 0.25" / 76.2mm x 6.4mm Rectangle Light Yellow From -80°C to +110°C (-112°F to +230°F)
Catalog No. 5000571048
Encompass_Preferred
Deep-Freeze permanent labels designed for specific use with high-profile 8-well PCR tube strips. Designed with vertical perforations along the tag, which allow each individual tube to be labeled securely. Ideal for long-term deep-freeze preservation in laboratory -80°C (-112°F) freezers and for dry ice transportation. They also withstand temperatures up to +110°C (+230°F) for safe use in PCR thermal cyclers. Labels can be printed with all major brands of thermal-transfer printers. Sets of 8 PCR labels
Catalog No. 50-005-71048 Supplier GA International Supplier No. FJT269C12LYE
Deep-Freeze permanent labels designed for specific use with high-profile 8-well PCR tube strips. Designed with vertical perforations along the tag, which allow each individual tube to be labeled securely. Ideal for long-term deep-freeze preservation in laboratory -80°C (-112°F) freezers and for dry ice transportation. They also withstand temperatures up to +110°C (+230°F) for safe use in PCR thermal cyclers. Labels can be printed with all major brands of thermal-transfer printers. Sets of 8 PCR labels