Learn More
GA International Deep-Freeze Labels for Inkjet Printers 1" x 1" / 25.4mm x 25.4mm Rectangle White From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000570997
Encompass_Preferred
Print eye-catching color logos, graphics, images, and data on our versatile inkjet-printable labels. These freezer-grade labels are designed to identify vials, tubes, plastic bags, freezer boxes, plates, and other containers. Ideal for storage in laboratory freezers (as low as -80°C/-112°F), as well as dry ice transportation, and can withstand exposure to alcohol, xylene, acetone, and other harsh chemicals (spray and incidental contact). Available in roll format, they can be printed with full-color inkjet printers; They can also be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.Compatible with PRIMERA LX500, LX500C, Kiaro QL-120, and Kiaro D printer models
Catalog No. 50-005-70997 Supplier GA International Supplier No. FIJA29C21WH
Print eye-catching color logos, graphics, images, and data on our versatile inkjet-printable labels. These freezer-grade labels are designed to identify vials, tubes, plastic bags, freezer boxes, plates, and other containers. Ideal for storage in laboratory freezers (as low as -80°C/-112°F), as well as dry ice transportation, and can withstand exposure to alcohol, xylene, acetone, and other harsh chemicals (spray and incidental contact). Available in roll format, they can be printed with full-color inkjet printers; They can also be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.Compatible with PRIMERA LX500, LX500C, Kiaro QL-120, and Kiaro D printer models