Learn More
GA International Deep-Freeze Removable Tape 0.5" x 50' / 12.7mm x 15m Green Apple 1" / 25.4mm From -80°C to +100°C (-112°F to +212°F)
Catalog No. NC2754916
Encompass_Preferred
Deep-freeze removable tape for sealing or identifying cryo and freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers (-20°C, -40°C, -80°C) and for dry ice transportation. Writable with our permanent cryo markers, glove-friendly, withstands dry ice, multiple freeze-thaw cycles, gamma irradiation, and other harsh laboratory environments. The tape should be applied at room temperature before storage in freezers. Not recommended for use in liquid nitrogen.
Catalog No. NC2754916 Supplier GA International Supplier No. TRM13C150GA
Deep-freeze removable tape for sealing or identifying cryo and freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers (-20°C, -40°C, -80°C) and for dry ice transportation. Writable with our permanent cryo markers, glove-friendly, withstands dry ice, multiple freeze-thaw cycles, gamma irradiation, and other harsh laboratory environments. The tape should be applied at room temperature before storage in freezers. Not recommended for use in liquid nitrogen.