Learn More
Grainger SUNEX Tools Impact Socket Set, 1/2 in. Drive, 6-Point, Deep, 29-Piece
Catalog No. 502777181
Encompass_Preferred
This 29-piece master set provides a comprehensive range of 1/2 in. drive SAE and metric deep impact sockets. Engineered for professional high-torque applications, these tools are constructed from durable chrome molybdenum alloy steel. The design features a 6-point configuration with radius corners to improve fastener engagement and prevent rounding.
  • 6-point design with radius corners improves engagement and reduces rounding.
  • Dual identification features laser-etched and permanent stamped markings.
  • Industrial-grade black phosphate finish provides superior corrosion resistance.
  • Includes adjustable metal storage rails with independent studs for organization.
  • Meets or exceeds ANSI and ASME performance standards.
  • Backed by a lifetime warranty covering defects in materials and workmanship.
Catalog No. 50-277-7181 Supplier Grainger Supplier No. 182K06
This 29-piece master set provides a comprehensive range of 1/2 in. drive SAE and metric deep impact sockets. Engineered for professional high-torque applications, these tools are constructed from durable chrome molybdenum alloy steel. The design features a 6-point configuration with radius corners to improve fastener engagement and prevent rounding.
  • 6-point design with radius corners improves engagement and reduces rounding.
  • Dual identification features laser-etched and permanent stamped markings.
  • Industrial-grade black phosphate finish provides superior corrosion resistance.
  • Includes adjustable metal storage rails with independent studs for organization.
  • Meets or exceeds ANSI and ASME performance standards.
  • Backed by a lifetime warranty covering defects in materials and workmanship.