Learn More
MSC Proto Deep Socket, 3/8 in, 11/32 in, 6-Point, 1 ea
Catalog No. 501920228
Encompass
This Proto deep hand socket is designed for various fastening applications, providing enhanced reach and grip in confined spaces. Made from durable alloy steel, it meets B107.1 standards for reliability and performance.
  • Features a 3/8 in drive size
  • Includes an 11/32 in socket size
  • Designed with a 6-point configuration
  • Overall length of 1/8 in
  • Chrome finish for corrosion resistance
  • Constructed from alloy steel for durability
  • Meets B107.1 standards
  • Not tether capable
Catalog No. 50-192-0228 Supplier MSC Supplier No. 80392202
This Proto deep hand socket is designed for various fastening applications, providing enhanced reach and grip in confined spaces. Made from durable alloy steel, it meets B107.1 standards for reliability and performance.
  • Features a 3/8 in drive size
  • Includes an 11/32 in socket size
  • Designed with a 6-point configuration
  • Overall length of 1/8 in
  • Chrome finish for corrosion resistance
  • Constructed from alloy steel for durability
  • Meets B107.1 standards
  • Not tether capable