Learn More
MSC Proto Deep Hand Socket, 3/8 Drive, 11 mm, 1 ea
Catalog No. 501920412
Encompass
The Proto J5011MH is a deep hand socket designed for general fastening applications. Featuring a 3/8 inch drive and an 11 mm, 6-Point opening, it provides efficient and reliable engagement with fasteners.
  • TorquePlus design minimizes fastener rounding and increases torque transfer
  • Exceeds ASME B107 specifications for performance and durability
  • Hardness rating of 47 to 51 HRC for long-lasting performance
  • Constructed from durable alloy steel
  • Chrome finish provides corrosion resistance
Catalog No. 50-192-0412 Supplier MSC Supplier No. 39934807
The Proto J5011MH is a deep hand socket designed for general fastening applications. Featuring a 3/8 inch drive and an 11 mm, 6-Point opening, it provides efficient and reliable engagement with fasteners.
  • TorquePlus design minimizes fastener rounding and increases torque transfer
  • Exceeds ASME B107 specifications for performance and durability
  • Hardness rating of 47 to 51 HRC for long-lasting performance
  • Constructed from durable alloy steel
  • Chrome finish provides corrosion resistance