Learn More
MSC Proto Deep Socket, 3/8 in Drive, 13mm, 6-Point, 1 ea
Catalog No. 501920414
Encompass
This Proto deep hand socket features a 3/8 inch drive and a 13mm, 6-point opening, engineered for optimal engagement with stubborn, damaged, or soft fasteners. Its specialized design ensures efficient power transfer while minimizing fastener wear.
  • Allows quick and easy socket engagement with fastener flats, not corners
  • Shifts contact area and stress away from fastener corners
  • Distributes contact stresses over a larger area of the socket, minimizing slippage
  • Minimizes fastener rounding and extends fastener life
  • Enables easier turning of worn, rounded, or rusted fasteners
Catalog No. 50-192-0414 Supplier MSC Supplier No. 39935143
This Proto deep hand socket features a 3/8 inch drive and a 13mm, 6-point opening, engineered for optimal engagement with stubborn, damaged, or soft fasteners. Its specialized design ensures efficient power transfer while minimizing fastener wear.
  • Allows quick and easy socket engagement with fastener flats, not corners
  • Shifts contact area and stress away from fastener corners
  • Distributes contact stresses over a larger area of the socket, minimizing slippage
  • Minimizes fastener rounding and extends fastener life
  • Enables easier turning of worn, rounded, or rusted fasteners