Learn More
CELLTREAT Scientific Products 81 Place Deep Storage Box for CF Cryogenic Vial, Polycarbonate, Non-sterile
SDP
Catalog No. NC1998689
Encompass_Preferred
Durable polycarbonate material designed to withstand a wide temperature range and is a reusable, autoclavable alternative to disposable boxes. Ventilation holes in the lids and bases prevent vacuum sealing in freezer. Storage Boxes are labeled with numeric markings for easy identification of individual positions. Safe to use at ultra-low temperatures. 100 place box features ridges along the sides to improve handling.
Catalog No. NC1998689 Supplier CELLTREAT Scientific Products Supplier No. 229944
Durable polycarbonate material designed to withstand a wide temperature range and is a reusable, autoclavable alternative to disposable boxes. Ventilation holes in the lids and bases prevent vacuum sealing in freezer. Storage Boxes are labeled with numeric markings for easy identification of individual positions. Safe to use at ultra-low temperatures. 100 place box features ridges along the sides to improve handling.