Learn More
New England Biolabs, Inc. Deep Vent DNA Polymerase – 1000 units
SDP
Catalog No. 50811678
Encompass
Deep Vent DNA Polymerase was the second high-fidelity thermophilic DNA polymerase available from New England Biolabs. The fidelity of Deep Vent DNA Polymerase is derived in part from an integral 3 5 proofreading exonuclease activity. Deep Vent is even more stable than Vent at temperatures of 95 to 100C.
Catalog No. 50-811-678 Supplier New England Biolabs, Inc. Supplier No. M0258L
Deep Vent DNA Polymerase was the second high-fidelity thermophilic DNA polymerase available from New England Biolabs. The fidelity of Deep Vent DNA Polymerase is derived in part from an integral 3 5 proofreading exonuclease activity. Deep Vent is even more stable than Vent at temperatures of 95 to 100C.