Learn More
Mascon Medical Mascon Medical™ Deep Well Plate, 96-Well, 2.0ml, 5EA/Bag, 10 Bag/Case
SDP
Supplier:  Mascon Medical FSDWP2
Encompass_Preferred
Mascon Medical™ 2.0ml Deep Well Plate is made of high-quality, medical-grade polypropylene with excellent chemical resistance. It is a reliable solution for sample storage, high-throughput screening, and low-binding experiments. It aligns with the international SBS standard and is suitable for automatic nucleic acid extraction and SBS automatic instruments. Best used for drug discovery, molecular biology, and genomic application. Works well with our Mascon Medical™ Sealing Film.Features:•Meets ISO13485 quality management system standard•Low retention•Low absorptionSpecifications:•Volume: 96-Well 2.0ml•Qty. / Case: 50•Color: Transparent•Shape: Round well, u-shaped bottom•Material: Polypropylene•Temperature Range: -80℃~120℃•Size: 127.3*85.3*45mm•Packaging Format: 5EA/Bag, 10Bags/Case
Catalog No. NC2458310
 Restricted Item