Learn More
LUNA NANOTECH INC Deep Well Plate Magnetic Rack
Catalog No. 502566969
Encompass_Preferred
- Designed to be used with different formats of 96-well deep well plates
- 24 magnets are positiont between the wells of the deep well plates for efficient magnetic separation
- Can be used for high throughput separation of up to 96 samples
- Housing material: PMMA/Acrylic (white)
- Magnet: N38 Neodymium (NdFeB)
Catalog No. 50-256-6969 Supplier LUNA NANOTECH INC Supplier No. MGRDWP
- Designed to be used with different formats of 96-well deep well plates
- 24 magnets are positiont between the wells of the deep well plates for efficient magnetic separation
- Can be used for high throughput separation of up to 96 samples
- Housing material: PMMA/Acrylic (white)
- Magnet: N38 Neodymium (NdFeB)