Learn More
Research Products International Corp Sealing Mats for Deep Well Plates, Non-Sterile, fits 96 Round Wells, 10 per Package
SDP
Catalog No. 501537649
Encompass
Sealing Mats are manufactured of durable, pierceable silicone. Designed to fit Deep Well Plates and preserve contents and are suitable for heat sealing. Sealing Mats can be rewashed and reused, but autoclaving is not recommended.
Catalog No. 50-153-7649 Supplier Research Products International Corp Supplier No. 162524
Sealing Mats are manufactured of durable, pierceable silicone. Designed to fit Deep Well Plates and preserve contents and are suitable for heat sealing. Sealing Mats can be rewashed and reused, but autoclaving is not recommended.