Learn More
Beckman Coulter DEEP WELL PLATE (STERILE)24/BX
Catalog No. NC9239209
Encompass
Plate, Deep-Well; Sterile; Biomek 96; 1mL/Well capacity; Natural; PP; Well Shape round; Available for storage or incubation; 24/CS
Catalog No. NC9239209 Supplier Beckman Coulter Supplier No. 267007
Plate, Deep-Well; Sterile; Biomek 96; 1mL/Well capacity; Natural; PP; Well Shape round; Available for storage or incubation; 24/CS