Learn More
GRANT INSTRUMENTS Thermoshaker for deepwell plates holds 1 plate, ambient +5° - +100°C, 250-1,400rpm
SDP
Numéro de catalogue. NC1546949
Encompass_Preferred
Grant TS-DW is a deep well plate Thermoshaker - designed for shaking and heating of deep well plates.A multisystem principle, used in design of the thermoshaker allows it to operate as 3 independent devices:Incubator.Plate shaker.Thermoshaker.Applications:Life science applications, molecular biology, cell biology lab, cell lysis, DNA isolation and purification, sample preparation for PCR, pellet re-suspension, or any other method where you have many samples that need mixing in deep well plates.
Numéro de catalogue. NC1546949 Fournisseur GRANT INSTRUMENTS Code fournisseur TSDW USA
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Grant TS-DW is a deep well plate Thermoshaker - designed for shaking and heating of deep well plates.A multisystem principle, used in design of the thermoshaker allows it to operate as 3 independent devices:Incubator.Plate shaker.Thermoshaker.Applications:Life science applications, molecular biology, cell biology lab, cell lysis, DNA isolation and purification, sample preparation for PCR, pellet re-suspension, or any other method where you have many samples that need mixing in deep well plates.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.