Learn More
MTC Bio Deep Well Plate and Tip Comb Combo pack 96 Square Well 2 2ml 44 2mm height V-bottom for Kingfisher Black Alphanumeric Printing PP Tip combs packed inside plates Sterile 10 bags of 5 sets 50/cs
Catalog No. 5000460174
Encompass_Preferred
Deep Well Plate and Tip Comb Combo pack 96 Square Well 2 2ml 44 2mm height V-bottom for Kingfisher Black Alphanumeric Printing PP Tip combs packed inside plates Sterile 10 bags of 5 sets 50/cs
Catalog No. 50-004-60174 Supplier MTC Bio Supplier No. D3096KFC
Add to Cart
Add to Cart
Deep Well Plate and Tip Comb Combo pack 96 Square Well 2 2ml 44 2mm height V-bottom for Kingfisher Black Alphanumeric Printing PP Tip combs packed inside plates Sterile 10 bags of 5 sets 50/cs