Learn More
Research Products International Corp Deep Well Plates, Sterile, 96 Square Round Wells, 2.0ml, 5 per Package
SDP
Catalog No. 501537646
Encompass
Deep Well Plates are ideal for sample collection, preparation and long-term storage. Raised rim on each well facilitates secure sealing with film or sealing mat. Plates allow optimum sample mixing and recovery through highly polished walls and offer reliable automation performance. Wells feature alphanumeric indexing molded in on the A-H and 1-12 axis with maximum centrifugation to 4,000 RCF.
Manufactured from virgin polypropylene (autoclavable) and are ultra-clear for easy visualization of samples. Plates are compatible with most liquid handling robots and SBS footprint meets ANSI/SLAS 1-2004. Stackable and flatness tested for reliable automation performance.
Catalog No. 50-153-7646 Supplier Research Products International Corp Supplier No. 162522S
Deep Well Plates are ideal for sample collection, preparation and long-term storage. Raised rim on each well facilitates secure sealing with film or sealing mat. Plates allow optimum sample mixing and recovery through highly polished walls and offer reliable automation performance. Wells feature alphanumeric indexing molded in on the A-H and 1-12 axis with maximum centrifugation to 4,000 RCF.
Manufactured from virgin polypropylene (autoclavable) and are ultra-clear for easy visualization of samples. Plates are compatible with most liquid handling robots and SBS footprint meets ANSI/SLAS 1-2004. Stackable and flatness tested for reliable automation performance.