Learn More
Labcon North America DEEP WELL PLATES WITH PRINTED
Catalog No. NC2008276
Encompass_Preferred
Deep Well Plates with Printed Grid | They include smooth inside surfaces to ensure all of your sample can be pipetted. Plates are designed to be manipulated by hand or by robots. Made from 100% virgin polypropylene and lot tested and certified to be free of RNase, DNase, and ATP.
Catalog No. NC2008276 Supplier Labcon North America Supplier No. 39085200009
Deep Well Plates with Printed Grid | They include smooth inside surfaces to ensure all of your sample can be pipetted. Plates are designed to be manipulated by hand or by robots. Made from 100% virgin polypropylene and lot tested and certified to be free of RNase, DNase, and ATP.