Learn More
Arcticwhite LLC 96 Square Deep Well, 2.2 ML/Well, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS (SBS) Standard Footprint, 2.2 ML Maximum, Polypropylene (PP), Medical-grade, Non-sterile, Conical (V-bottom), Storage Down to -86 Deg C, 50 Deep Well Plates per Case
SDP
Catalog No. NC9640167
Encompass
96 Square Deep Well, 2.2 ML/Well, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS (SBS) Standard Footprint, 2.2 ML Maximum, Polypropylene (PP), Medical-grade, Non-sterile, Conical (V-bottom), Storage Down to -86 Deg C, 50 Deep Well Plates per Case. This is a 96-well deep well microplate for sample storage, automated liquid handling, nucleic acid purification, and cell culture. Working volume - 2 mL. Material - Polypropylene (PP), medical-grade. Non-sterile. DNase-free and RNase-free. Thermal range - Storage down to -86 deg C. Supplied by ArcticWhite as 50 deep well plates per case.
Catalog No. NC9640167 Supplier Arcticwhite LLC Supplier No. AWLS219009
96 Square Deep Well, 2.2 ML/Well, Polypropylene, Conical Bottom, 96-well, ANSI/SLAS (SBS) Standard Footprint, 2.2 ML Maximum, Polypropylene (PP), Medical-grade, Non-sterile, Conical (V-bottom), Storage Down to -86 Deg C, 50 Deep Well Plates per Case. This is a 96-well deep well microplate for sample storage, automated liquid handling, nucleic acid purification, and cell culture. Working volume - 2 mL. Material - Polypropylene (PP), medical-grade. Non-sterile. DNase-free and RNase-free. Thermal range - Storage down to -86 deg C. Supplied by ArcticWhite as 50 deep well plates per case.