Learn More
Beckman Coulter 96-Round Deep Well Plates Non-Sterile, 24/cs
Catalog No. NC1482478 Shop All Beckman Coulter Products
Encompass
Ideal for sample collections, compound preparation, combinatorial chemistry, high throughput screening, nucleic acid purification, bacterial culture growth and plate replication. Completely automation-friendly with 1 mL per well volume. Also available in polypropylene and pre-sterile formats.
Catalog No. NC1482478 Supplier Beckman Coulter Supplier No. 267001
Ideal for sample collections, compound preparation, combinatorial chemistry, high throughput screening, nucleic acid purification, bacterial culture growth and plate replication. Completely automation-friendly with 1 mL per well volume. Also available in polypropylene and pre-sterile formats.