Learn More
Sigma Aldrich Fine Chemicals Biosciences Deepwell 96 well plate pol
Catalog No. 502905291
Encompass
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays enzyme or receptor assays or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.
Catalog No. 50-290-5291 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAF37860000124EA
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays enzyme or receptor assays or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.