Learn More
Waters Corp Deepwell Magnet, Automated Moving Shelf, DNA Purification
Catalog No. 502390871 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Deepwell Magnet+ provides a simple and reliable method for purifying genomic, plasmid, and mitochondrial DNA using magnetic beads. It features an automated moving shelf that strategically positions magnets near the bottom sides of consumables to optimize bead pelleting.
  • Ensures simple and reliable purification of genomic, plasmid, and mitochondrial DNA using magnetic beads.
  • Automated moving shelf for strategic magnet positioning to optimize bead pelleting.
Catalog No. 50-239-0871 Supplier Waters Corp Supplier No. 176004854
Add to Cart
Add to Cart
Request a Quote
The Deepwell Magnet+ provides a simple and reliable method for purifying genomic, plasmid, and mitochondrial DNA using magnetic beads. It features an automated moving shelf that strategically positions magnets near the bottom sides of consumables to optimize bead pelleting.
  • Ensures simple and reliable purification of genomic, plasmid, and mitochondrial DNA using magnetic beads.
  • Automated moving shelf for strategic magnet positioning to optimize bead pelleting.