Learn More
Waters Corp Waters Storage Plate Domino, Machine-Readable, Andrew+ Compatible, QR/ARUCO Codes
Supplier:  Waters Corp 186009596
Encompass_Preferred
Dominos are machine-readable consumable holders that create the interface between a variety of different consumable types and the Andrew+ pipetting robot to enable seamless automation of many different laboratory workflows. Equipped with QR codes housing critical labware information and ARUCO codes for spatial positioning, Dominos are an indispensable part of any Andrew+ automation configuration.
  • Machine-readable consumable holders
  • Creates an interface between various consumable types and the Andrew+ pipetting robot
  • Enables seamless automation of laboratory workflows
  • Equipped with QR codes for critical labware information
  • Equipped with ARUCO codes for spatial positioning
Catalog No. 50-226-9319