Learn More
Waters Corp Waters Corporation Deepwell Plate Fraction Collector Rack Kit, for Fraction Collectors
Catalog No. 50791623 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This deepwell plate fraction collector rack kit is designed for use with fraction collectors, specifically enabling the collection of fractions into deepwell plates during various chromatographic applications. It is part of the range of service parts and kits.
  • Facilitates fraction collection into deepwell plates.
  • Compatible with a range of fraction collectors.
  • Supports diverse chromatographic applications, including HPLC and LC-MS.
  • Offers flexible collection of various fraction sizes.
Catalog No. 50-791-623 Supplier Waters Corp Supplier No. 205000134
This deepwell plate fraction collector rack kit is designed for use with fraction collectors, specifically enabling the collection of fractions into deepwell plates during various chromatographic applications. It is part of the range of service parts and kits.
  • Facilitates fraction collection into deepwell plates.
  • Compatible with a range of fraction collectors.
  • Supports diverse chromatographic applications, including HPLC and LC-MS.
  • Offers flexible collection of various fraction sizes.