Learn More
Kuhner Shaker Inc DEEPWELL SANDWICH COVER 96 WEL
Catalog No. NC1102631
Encompass
Sandwich cover for all types of 96 deepwell MTPs, (with spongy silicone, 1.5 mm holes)
Catalog No. NC1102631 Supplier Kuhner Shaker Inc Supplier No. SMCR1296
Sandwich cover for all types of 96 deepwell MTPs, (with spongy silicone, 1.5 mm holes)