Learn More
American Research Products Inc Deer Insulin like growth facto
Catalog No. 5000329789
Encompass
Deer Insulin-like growth factors 1 IGF-1 ELISA Kit from CUSABIO Detection Range 1 ng/mL-200 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29789 Supplier American Research Products Inc Supplier No. CSBE12644DE
Deer Insulin-like growth factors 1 IGF-1 ELISA Kit from CUSABIO Detection Range 1 ng/mL-200 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive