Learn More
Apexbio Technology LLC DEET 134-62-3 1g
SDP
Catalog No. 5000089220
Encompass_Preferred
DEET (134-62-3) is a small-molecule antagonist targeting insect odorant receptors It is designed to disrupt molecular interactions within chemosensory pathways thereby inhibiting insect attraction and biting behavior DEET exerts its biological activity primarily through interference with insect odorant receptors In in vitro and behavioral assays DEET demonstrates inhibitory activity with variable IC50 values reported for receptor modulation and insect behavioral responses Based on these pharmacological properties DEET holds research potential in evaluating insect chemoreception and vector-host interactions relevant to the transmission of pathogens such as Zika virus malaria parasites West Nile virus Lyme disease-causing bacteria and Rocky Mountain spotted fever agents
Catalog No. 50-000-89220 Supplier Apexbio Technology LLC Supplier No. C67501000
DEET (134-62-3) is a small-molecule antagonist targeting insect odorant receptors It is designed to disrupt molecular interactions within chemosensory pathways thereby inhibiting insect attraction and biting behavior DEET exerts its biological activity primarily through interference with insect odorant receptors In in vitro and behavioral assays DEET demonstrates inhibitory activity with variable IC50 values reported for receptor modulation and insect behavioral responses Based on these pharmacological properties DEET holds research potential in evaluating insect chemoreception and vector-host interactions relevant to the transmission of pathogens such as Zika virus malaria parasites West Nile virus Lyme disease-causing bacteria and Rocky Mountain spotted fever agents