Learn More
Medchemexpress LLC Defactinib | 1073154-85-4 | 99.7% | 200 MG
SDP
Catalog No. 5000230351
Encompass_Preferred
Defactinib is an orally bioavailable small-molecule focal adhesion kinase (FAK) inhibitor used in research to study antiangiogenic and antineoplastic mechanisms. Supplied as a white to off-white solid with high reported purity, it is commonly offered in multiple research pack sizes and solution formats for cellular assays.
  • Orally bioavailable FAK inhibitor
  • Inhibits FAK phosphorylation and downstream signaling pathways
  • High reported purity (~99.7%) and supplied as a solid
  • Available in multiple research pack sizes and DMSO solutions
  • CAS number 1073154-85-4 and MFCD25977806 for identification
Catalog No. 50-002-30351 Supplier Medchemexpress LLC Supplier No. HY12289200MG
Defactinib is an orally bioavailable small-molecule focal adhesion kinase (FAK) inhibitor used in research to study antiangiogenic and antineoplastic mechanisms. Supplied as a white to off-white solid with high reported purity, it is commonly offered in multiple research pack sizes and solution formats for cellular assays.
  • Orally bioavailable FAK inhibitor
  • Inhibits FAK phosphorylation and downstream signaling pathways
  • High reported purity (~99.7%) and supplied as a solid
  • Available in multiple research pack sizes and DMSO solutions
  • CAS number 1073154-85-4 and MFCD25977806 for identification