Learn More
Medchemexpress LLC HY-12289 10mg Medchemexpress, Defactinib CAS:1073154-85-4 Purity:>98%
SDP
Catalog No. 502028870
Encompass_Preferred
Medchemexpress, HY-12289 10mg Defactinib CAS:1073154-85-4 Defactinib (VS-6063; PF-04554878) is a novel FAK inhibitor with potential antiangiogenic and antineoplastic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8870 Supplier Medchemexpress LLC Supplier No. HY1228910MG
Medchemexpress, HY-12289 10mg Defactinib CAS:1073154-85-4 Defactinib (VS-6063; PF-04554878) is a novel FAK inhibitor with potential antiangiogenic and antineoplastic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.