Learn More
Medchemexpress LLC HY-12289A 5mg Medchemexpress, Defactinib (hydrochloride) CAS:1073160-26-5 Purity:>98%
SDP
Catalog No. 502080998
Encompass_Preferred
Medchemexpress, HY-12289A 5mg Defactinib (hydrochloride) CAS:1073160-26-5 Defactinib hydrochloride (VS-6063 hydrochloride; PF 04554878 hydrochloride) is a novel FAK inhibitor, which inhibits FAK phosphorylation at the Tyr397 site in a time- and dose-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0998 Supplier Medchemexpress LLC Supplier No. HY12289A5MG
Medchemexpress, HY-12289A 5mg Defactinib (hydrochloride) CAS:1073160-26-5 Defactinib hydrochloride (VS-6063 hydrochloride; PF 04554878 hydrochloride) is a novel FAK inhibitor, which inhibits FAK phosphorylation at the Tyr397 site in a time- and dose-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.