Learn More
Medchemexpress LLC Defactinib hydrochloride | 1073160-26-5 | 99.4% | 1 ML
SDP
Catalog No. 5000232112
Encompass_Preferred
Defactinib hydrochloride is a small-molecule focal adhesion kinase (FAK) inhibitor that suppresses FAK Tyr397 phosphorylation in a time- and dose-dependent manner. It is supplied as the hydrochloride salt and is available as a 10 mM solution in DMSO (1 mL) or as solid samples for biochemical and cellular research.
  • Inhibits FAK phosphorylation at Tyr397
  • Supplied as hydrochloride salt for improved solubility
  • Available as 10 mM solution in DMSO (1 mL) and as solid samples
  • High purity suitable for research assays
  • Useful for studies of cell migration, invasion, and signaling
Catalog No. 50-002-32112 Supplier Medchemexpress LLC Supplier No. HY12289A10MM 1ML
Defactinib hydrochloride is a small-molecule focal adhesion kinase (FAK) inhibitor that suppresses FAK Tyr397 phosphorylation in a time- and dose-dependent manner. It is supplied as the hydrochloride salt and is available as a 10 mM solution in DMSO (1 mL) or as solid samples for biochemical and cellular research.
  • Inhibits FAK phosphorylation at Tyr397
  • Supplied as hydrochloride salt for improved solubility
  • Available as 10 mM solution in DMSO (1 mL) and as solid samples
  • High purity suitable for research assays
  • Useful for studies of cell migration, invasion, and signaling