Learn More
Medchemexpress LLC Defactinib hydrochloride | 1073160-26-5 | 99.4% | 10 MG
SDP
Catalog No. 5000233775
Encompass_Preferred
Defactinib hydrochloride is the hydrochloride salt of defactinib, a small-molecule focal adhesion kinase (FAK) inhibitor. It inhibits FAK phosphorylation at Tyr397 in a time- and dose-dependent manner and is used in preclinical research to study tumor biology and signaling pathways.
  • Inhibits FAK phosphorylation at Tyr397 in a time- and dose-dependent manner.
  • Hydrochloride salt supplied as a white to off-white solid.
  • High chemical purity suitable for biochemical and cellular assays.
  • Available in multiple solid sizes and DMSO stock solutions for flexible dosing.
  • Used for research into cancer signaling and pharmacology applications.
Catalog No. 50-002-33775 Supplier Medchemexpress LLC Supplier No. HY12289A10MG
Defactinib hydrochloride is the hydrochloride salt of defactinib, a small-molecule focal adhesion kinase (FAK) inhibitor. It inhibits FAK phosphorylation at Tyr397 in a time- and dose-dependent manner and is used in preclinical research to study tumor biology and signaling pathways.
  • Inhibits FAK phosphorylation at Tyr397 in a time- and dose-dependent manner.
  • Hydrochloride salt supplied as a white to off-white solid.
  • High chemical purity suitable for biochemical and cellular assays.
  • Available in multiple solid sizes and DMSO stock solutions for flexible dosing.
  • Used for research into cancer signaling and pharmacology applications.