Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Defactinib hydrochloride | 1073160-26-5 | 99.4% | 50 MG
SDP

Catalog No. 5000230520
Encompass_Preferred

Defactinib hydrochloride is the hydrochloride salt of defactinib (VS-6063; PF-04554878), a small-molecule focal adhesion kinase (FAK) inhibitor used in research to study FAK signaling, tumor cell motility, and antineoplastic and antiangiogenic mechanisms.

  • Inhibits FAK phosphorylation at Tyr397 in a time- and dose-dependent manner.
  • Hydrochloride salt form, off-white powder suitable for research use.
  • Molecular formula C20H22ClF3N8O3S; molecular weight 546.95 g/mol.
  • Reported purity 99.35% with available certificate of analysis.
  • Available in multiple pack sizes, including 50 mg.
  • Supporting documents available: data sheet, COA, and SDS.

Catalog No. 50-002-30520 Supplier Medchemexpress LLC Supplier No. HY12289A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Defactinib hydrochloride is the hydrochloride salt of defactinib (VS-6063; PF-04554878), a small-molecule focal adhesion kinase (FAK) inhibitor used in research to study FAK signaling, tumor cell motility, and antineoplastic and antiangiogenic mechanisms.

  • Inhibits FAK phosphorylation at Tyr397 in a time- and dose-dependent manner.
  • Hydrochloride salt form, off-white powder suitable for research use.
  • Molecular formula C20H22ClF3N8O3S; molecular weight 546.95 g/mol.
  • Reported purity 99.35% with available certificate of analysis.
  • Available in multiple pack sizes, including 50 mg.
  • Supporting documents available: data sheet, COA, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.