Learn More
Aviva Systems Biology Corp DEFB4A Antibody - N-terminal region (ARP60172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DEFB4A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DEFB4A. Alternate names: DEFB-2, DEFB102, DEFB2, DEFB4, HBD-2, SAP1, BD-2ApplicationWB. Species reactivity: Human
Catalog No. 50-286-969