Learn More
OHAUS™ Defender 6000 Extreme Washdown Scale Indicator
Catalog No. 01919429 Shop All OHAUS Products
Change view
Click to view available options
Construction:
Polycarbonate, Stainless Steel (316)
Stainless Steel (316)
Model:
i-DT61PW AM
i-DT61PW AM Front Installation
i-DT61XWE AM
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Construction Model
01-919-429 Polycarbonate, Stainless Steel (316) i-DT61PW AM
01-919-430 Polycarbonate, Stainless Steel (316) i-DT61PW AM Front Installation
01-919-431 Stainless Steel (316) i-DT61XWE AM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-919-429 Supplier OHAUS™ Supplier No. 30575583
Add to Cart
Add to Cart
The Defender 6000 Series Extreme Washdown Indicator is specifically designed for food, chemical and pharmaceutical applications including production, processing, and packaging.
  • Applications: Weighing, Counting with APW optimization, Percent Weighing, Checkweighing, Checkcounting, Dynamic Weighing/Display Hold
  • Display: 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
  • Operation: 6 D (LR20) Batteries (Included)
  • Communication: Easy access IR communication kit (available as an accessory); GLP/GMP data output with real-time clock
  • Construction: IP68/IP69K food grade polycarbonate housing and 316 stainless steel mounting bracket
  • Design Features: 3-color check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare
TRUSTED_SUSTAINABILITY
Specifications
Battery Life Up to 1,600 hours continuous use with backlight off
Construction Polycarbonate, Stainless Steel (316)
Display Type Backlit LCD
Model i-DT61PW AM
Protection Class IP68 immersion/IP69K washdown
Type Bench Scales
Legal for Trade Class III/IIIL/IIIHD