Learn More
Cayman Chemical Defensamide, Purity: ≥98%, CAS: 1104874-94-3, 50 mg
SDP
Catalog No. 502917442
Encompass_Preferred
An SPHK1 activator; increases S1P levels in primary human keratinocytes but does not affect S1P lyase activity in the same cells at 100 µM; increases protein levels of LL-37 in primary human keratinocytes at 100 µM; inhibits invasion of S. aureus into murine skin ex vivo; has been used as an intermediate in the synthesis of sinuxylamide A as well as compounds with anticancer and antibacterial activities
Catalog No. 50-291-7442 Supplier Cayman Chemical Supplier No. 3731850 MG
An SPHK1 activator; increases S1P levels in primary human keratinocytes but does not affect S1P lyase activity in the same cells at 100 µM; increases protein levels of LL-37 in primary human keratinocytes at 100 µM; inhibits invasion of S. aureus into murine skin ex vivo; has been used as an intermediate in the synthesis of sinuxylamide A as well as compounds with anticancer and antibacterial activities