Learn More
Carbosynth LLC. α-Defensin-5 (Human) (0.1mg vial), 1ea, Disulfide bonds: (Disulfide bonds between Cys3-Cys31, Cys5-Cys20, and Cys10-Cys30), MW: 3582.1, Molecular formula: C144H238N50O45S6
SDP
Supplier:  Carbosynth LLC. PDF4415S0.1MG
Encompass_Preferred
Synonyms: Ala-Thr-Cys-Tyr-Cys-Arg-Thr-Gly-Arg-Cys-Ala-Thr-Arg-Glu-Ser-Leu-Ser-Gly-Val-Cys-Glu-Ile-Ser-Gly-Arg-Leu-Tyr-Arg-Leu-Cys-Cys-Arg, HD-5, Human Defensin-5, alpha-Defensin-5, DEFA5, H-ATCYCRTGRCATRESLSGVCEISGRLYRLCCR-OH, Application: Antimicrobial Peptide in Paneth Cells, Storage Temperature: -20°C, References: T. Ganz, M.E. Selsted, D. Szklarek, S.S.L. Harwig, K. Daher, D.F. Bainton, and R.I. Lehrer, J. Clin. Invest., 76, 1427 (1985). (Original; Isolation of HNP 1-3), M.E. Selsted, S.S.L. Harwig, T. Ganz, J.W. Schilling, and R.I. Lehrer, J. Clin. Invest., 76, 1436 (1985). (Original; Structure of HNP 1-3), X.-M. Fang, Q. Shu, Q.-X. Chen, M. Book, H.-G. Sahl, A. Hoeft, and F. Stuber, Eur. J. Clin. Invest., 33, 82 (2003). (Histochem.; Regulation of Expression)
Catalog No. 50-168-6419