Learn More
Bachem Defensin HNP-1 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4025473.0100
Encompass_Preferred
Defensin HNP-1 (human) Trifluoroacetate, H-Ala-Cys-Tyr-Cys-Arg-Ile-Pro-Ala-Cys-Ile-Ala-Gly-Glu-Arg-Arg-Tyr-Gly-Thr-Cys-Ile-Tyr-Gln-Gly-Arg-Leu-Trp-Ala-Phe-Cys-Cys-OH, 148093-65-6, C150H222N44O38S6, Mr 3442.08, 0.1mg. HNP-1, Human Neutrophil Peptide-1.
Catalog No. 50-260-257