Learn More
CPC Scientific H-Cys-Tyr-Cys-Arg-Ile-Pro-Ala-Cys-Ile-Ala-Gly-Glu-Arg-Arg-Tyr-Gly-Thr-Cys-Ile-Tyr-Gln-Gly-Arg-Leu-Trp-Ala-Phe-Cys-Cys-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific DEFS001A
Encompass
SEQUENCE: H-Cys-Tyr-Cys-Arg-Ile-Pro-Ala-Cys-Ile-Ala-Gly-Glu-Arg-Arg-Tyr-Gly-Thr-Cys-Ile-Tyr-Gln-Gly-Arg-Leu-Trp-Ala-Phe-Cys-Cys-OH (trifluoroacetate salt)(Cys1 and 29 bridge, Cys3 and 18 bridge, Cys8 and 28 bridge)ONE-LETTER SEQUENCE: CYCRIPACIAGERRYGTCIYQGRLWAFCC (C1&C29 bridge, C3&C18 bridge, C8&C28 bridge)MOLECULAR FORMULA: C147H217N43O37S6MOLECULAR WEIGHT: 3370.95STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [99287-07-7]SYNONYMS: -Defensin-2 (Human), alpha-Defensin-2 (Human), Defensin HNP-2 humanRESEARCH AREA: AntimicrobialREFERENCES:M.E. Selsted, S.S.L. Harwig, T. Ganz, J.W. Schilling, and R.I. Lehrer, J. Clin. Invest., 76,1436 (1985). (Original)T. Ganz, M.E. Selsted, D. Szklarek, S.S.L. Harwig, K. Daher, D.F. Bainton, and R.I. Lehrer, J. Clin. Invest., 76,1427 (1985).(First Isolation)F.T. Lundy, D.F. Orr, C. Shaw, P.-J. Lamey, and G.J. Linden, Oral Oncol., 40, 139 (2004).C.B. Buck, P.M. Day, C.D. Thompson, J. Lubkowski, W. Lu, D.R. Lowy, a
Catalog No. 50-210-9184