Learn More
Enzo Life Sciences Defensins (human neutrophils) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5140050
Encompass_Preferred
Α-defensins purified from human neutrophils. Defensins are short peptides secreted by neutrophils and epithelial cells as proproteins and activated by proteolysis. They have antibacterial, antiviral, and antifungal properties, and they promote immune response. Can be used in tissue culture or in vivo. Alternative name: HNPs, Human neutrophil peptides. Purity: ≥95%. Long Term Storage: -80°C.
Catalog No. 50-201-2059